missing translation for 'onlineSavingsMsg'
Learn More

Gamitrinib TPP hexafluorophosphate, MedChemExpress

Product Code. 16456417
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456417 5 mg 5mg
16450969 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456417 Supplier Medchem Express Supplier No. HY102007A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gamitrinib TPP hexafluorophosphate is a Gamitrinib (GA) mitochondrial matrix inhibitor. Gamitrinib TPP hexafluorophosphate is a mitochondrial targeted HSP90 inhibitor with anti-cancer activity.

Gamitrinib TPP hexafluorophosphate is a Gamitrinib (GA) mitochondrial matrix inhibitor. Gamitrinib TPP hexafluorophosphate is a mitochondrial targeted HSP90 inhibitor with anti-cancer activity.

Specifications

Chemical Name or Material Gamitrinib TPP hexafluorophosphate
Physical Form Solid
CAS 1131626-47-5
Color Pale Purple
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C52H65F6N3O8P2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (48.26 mM; Need ultrasonic)
SMILES F[P-](F)(F)(F)(F)F.O=C(C=C1NC(/C(C)=C/C=C/[C@H](OC)[C@@H](OC(N)=O)/C(C)=C/[C@H](C)[C@H]2O)=O)C(NCCCCCC[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)=C(C[C@H](C[C@@H]2OC)C)C1=O
Molecular Weight (g/mol) 1036.03
Formula Weight 1036.03
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.