missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamabufotalin, MedChemExpress
Description
Gamabufotalin (Gamabufagin), a main active compound isolated from Chinese medicine Chansu, has been shown to strongly inhibit cancer cell growth and inflammatory response. Gamabufotalin could inhibite angiogenesis by inhibiting the activation of VEGFR-2 signaling pathways.
Specifications
Specifications
| Chemical Name or Material | Gamabufotalin |
| CAS | 465-11-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H34O5 |
| Quantity | 5 mg |
| Synonym | Gamabufagin |
| Solubility Information | DMSO : 50 mg/mL (124.22 mM; Need ultrasonic) |
| SMILES | C[C@]([C@@H](C(C=C1)=COC1=O)CC2)(C[C@@H](O)[C@@]3([H])[C@@]4([H])CC[C@@]5([H])[C@@]3(CC[C@H](O)C5)C)[C@]24O |
| Molecular Weight (g/mol) | 402.52 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?