missing translation for 'onlineSavingsMsg'
Learn More

Galvinoxyl free radical, 98%, Thermo Scientific Chemicals

Product Code. 10439030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10439030 5 g Glass bottle 5g
10774231 1 g Glass bottle 1g
10095753 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10439030 Supplier Thermo Scientific Acros Supplier No. 119850050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Polymerization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2370-18-5
Molecular Formula C29H42O2
Molecular Weight (g/mol) 422.65
MDL Number MFCD00001616
InChI Key HRLDHROCDBYHAU-UHFFFAOYSA-N
Synonym 2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical
IUPAC Name 2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C

Specifications

Melting Point 156°C to 160°C
Color Blue to Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 08, IV, 1379
Fieser 01,409
Merck Index 15, 4387
Formula Weight 421.64
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Galvinoxyl free radical
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.