Learn More
Emma Safety Footwear Galus Safety Shoes
Product Code. 15978751
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15978751 37 1 pair
15988751 38 1 pair
15998751 39 1 pair
15908761 40 1 pair
15918761 41 1 pair
15928761 42 1 pair
15938761 43 1 pair
15948761 44 1 pair
15958761 45 1 pair
15968761 46 1 pair
15978761 47 1 pair
15988761 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15978751 missing translation for 'mfr' Emma Safety Footwear missing translation for 'supplierNo' MM58086037
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
Insulated with fine quality sheepskin to keep feet warm on the coldest of conditions. Nitrile rubber outsole won't yield if the temperature rises up to 300 degrees on the outside.
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Fur-lined and insulated for warmth
Specifikationer
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Zip
Color Black
Design Galus
Liner Material Fur
Material Leather
Size 37
Toe Material Steel
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.