missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Galunisertib
GREENER_CHOICE

Product Code. p-7166833
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18777976

Brand: Tocris Bioscience™ 6956/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ALK4 and ALK5 (TGFβRI) inhibitor

Galunisertib is an ALK4 and ALK5 (TGFβRI) inhibitor (IC50 values are 77.7 and 172 nM, respectively). Also inhibits a range of other kinases with submicromolar IC50s including MINK, TGFβRII, ALK6 and ACVR2B. Inhibits TGFβ-induced SMAD phosphorylation in cancer cell lines in vitro and in vivo. Also reverses TGFβ-mediated suppression of T cell proliferation. Prevents TGFβ-mediated EMT and cell migration in vitro and exhibits antitumor efficacy in vivo. Used as component of growth media for urothelial organoids. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[5,6-Dihydro-2-(6-methyl-2-pyridinyl)-4 H-pyrrolo[1,2-b]pyrazol-3-yl]-6-quinolinecarboxamide
CAS 700874-72-2
Quantity 5 mg
Target TGF-beta Receptor Inhibitors
Molecular Formula C22H19N5O
Formula Weight 369.43
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.