missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GALR3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA528747

Product Code. 13442867

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse eye, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The neuropeptide galanin modulates a variety of physiologic processes including cognition/memory, sensory/pain processing, hormone secretion, and feeding behavior. The human galanin receptors are G protein-coupled receptors that functionally couple to their intracellular effector through distinct signaling pathways. GALR3 is found in many tissues and may be expressed as 1.4-, 2.4-, and 5-kb transcripts. GALR3 has been reported in brain, testis, adrenal gland, and pancreas. An EST has been isolated from an ovarian thecoma fibroma library.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GALR3
Polyclonal
Unconjugated
GALR3
Gal receptor-3; Gal3 receptor; GAL3-R; Galanin r3 receptor; galanin receptor 3; galanin receptor type 3; galanin receptor, family member 3; Galnr3; Galr3; Gal-r3; GALR-3; OTTHUMP00000028820
Rabbit
Antigen affinity chromatography
RUO
14429, 29235, 8484
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.72 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O60755, O88626, O88853
GALR3
Synthetic peptide encompassing a sequence within the Extracellular domain of human GALR3. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.