missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALR3 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577442
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The neuropeptide galanin modulates a variety of physiologic processes including cognition/memory, sensory/pain processing, hormone secretion, and feeding behavior. The human galanin receptors are G protein-coupled receptors that functionally couple to their intracellular effector through distinct signaling pathways. GALR3 is found in many tissues and may be expressed as 1.4-, 2.4-, and 5-kb transcripts. GALR3 has been reported in brain, testis, adrenal gland, and pancreas. An EST has been isolated from an ovarian thecoma fibroma library.
Specifications
| GALR3 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Galr3 | |
| GALR3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14429, 29235 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.7 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O88626, O88853 | |
| Galr3 | |
| Peptide SYYGTVRYGALELC, corresponding to amino acid residues 159-172 of mouse Galanin receptor type 3 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction