missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALNTL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556616
This item is not returnable.
View return policy
Description
Immunogen sequence: DKHLVKSAEP GEQQTFPLGL GDGQFYSWTD GLRRKDWHDY ESIQKEAMRS GKGEHGKPYP LTEEDHDDSA YRENGFNIFV SN Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 37%.
Specifications
| GALNTL6 | |
| Polyclonal | |
| Unconjugated | |
| GALNTL6 | |
| GalNAc transferase 17; galNAc-T17; GALNACT20; GalNAc-T6L; GALNT17; GALNTL6; GaNTase 17; polypeptide GalNAc transferase 17; polypeptide GalNAc transferase-like 6; polypeptide N-acetylgalactosaminyltransferase-like 6; pp-GaNTase 17; protein-UDP acetylgalactosaminyltransferase 17; Putative polypeptide N-acetylgalactosaminyltransferase 17; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 17; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 20; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 442117 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q49A17 | |
| GALNTL6 | |
| Recombinant protein corresponding to Human GALNTL6. Recombinant protein control fragment (Product #RP-95006). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction