Learn More
Invitrogen™ GALNTL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140132
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: WVTNYIASVY VRGQEPAPDK KLEEDKGDTL KIIERLDHLE NVIKQHIQEA PAKPEEAEAE PFTDSSLFAH W Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 100%.
Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
Specifications
| GALNTL4 | |
| Polyclonal | |
| Unconjugated | |
| GALNT18 | |
| 2900011G21Rik; BC024988; GalNAc-T15; GALNACT18; GalNAc-T18; GalNAc-T-like protein 4; GalNAc-transferase 18; GALNT15; GALNT18; Galntl4; LOW QUALITY PROTEIN: polypeptide N-acetylgalactosaminyltransferase 18; mpp-GalNAc-T18(O18); polypeptide GalNAc transferase 18; Polypeptide GalNAc transferase-like protein 4; polypeptide N-acetylgalactosaminyltransferase 18; polypeptide N-acetylgalactosaminyltransferase-like 4; polypeptide N-acetylgalactosaminyltransferase-like protein 4; pp-GaNTase-like protein 4; protein-UDP acetylgalactosaminyltransferase-like protein 4; putative polypeptide N-acetylgalactosaminyltransferase-like protein 4; RGD1309623; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 374378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P9A2 | |
| GALNT18 | |
| Recombinant protein corresponding to Human GALNT18. Recombinant protein control fragment (Product #RP-109218). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.