missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALNT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526975
This item is not returnable.
View return policy
Description
GALNT5 Polyclonal Antibody for Flow, IHC (P), Western Blot
GALNT5 can catalyze the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. GALNT5 has activity toward EA2 peptide substrate, but it has a weak activity toward Muc2 or Muc1b substrates.
Specifications
| GALNT5 | |
| Polyclonal | |
| Unconjugated | |
| GALNT5 | |
| GALNACT5; GalNAc-T5; GALNT5; Polypeptide GalNAc transferase 5; polypeptide N-acetylgalactosaminyltransferase 5; pp-GaNTase 5; protein-UDP acetylgalactosaminyltransferase 5; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 5; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5 (GalNAc-T5) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11227 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q7Z7M9 | |
| GALNT5 | |
| KLH conjugated synthetic peptide between 24-53 amino acids from the N-terminal region of human GALNT5. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction