Learn More
Invitrogen™ GALNT16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140171
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QRAGRRSEQL REDRTIPLIV TGTPSKGFDE KAYLSAKQLK AGEDPYRQHA FNQLESDKLS PDRPIRDTRH YSCPS Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 96%.
GALNTL1 belongs to the glycosyltransferase 2 family, GalNAc-T subfamily. It contains 1 ricin B-type lectin domain. GALNTL1 may catalyze the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
Specifications
| GALNT16 | |
| Polyclonal | |
| Unconjugated | |
| GALNT16 | |
| 5730405L21Rik; AI415388; GALNACT16; galNAc-T16; GalNAc-T-like protein 1; GALNT16; GALNTL1; KIAA1130; MGC141855; mpp-GalNAc-T16; Polypeptide GalNAc transferase 16; polypeptide GalNAc transferase-like protein 1; Polypeptide N-acetylgalactosaminyltransferase 16; polypeptide N-acetylgalactosaminyltransferase-like 1; polypeptide N-acetylgalactosaminyltransferase-like protein 1; pp-GaNTase-like protein 1; protein-UDP acetylgalactosaminyltransferase-like protein 1; putative polypeptide N-acetylgalactosaminyltransferase-like protein 1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 1; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57452 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N428 | |
| GALNT16 | |
| Recombinant protein corresponding to Human GALNT16. Recombinant protein control fragment (Product #RP-109470). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.