missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GALNT15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15791481 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15791481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15791481 Supplier Invitrogen™ Supplier No. PA553548

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GGSVEILPCS RVGHIYQNQD SHSPLDQEAT LRNRVRIAET WLGSFKETFY KHSPEAFSLS KAEKPDCMER LQLQRRLGCR TFHWFLANVY PELYPSEPRP SFSGKLHNTG LG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 61%.

Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Although it displays a much weaker activity toward all substrates tested compared to GALNT2, it is able to transfer up to seven GalNAc residues to the Muc5AC peptide, suggesting that it can fill vicinal Thr/Ser residues in cooperation with other GALNT proteins. Prefers Muc1a as substrate. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT15
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT15
Gene Accession No. Q8N3T1
Gene Alias 4631401E18Rik; GALNACT15; GalNAc-T-like protein 2; GALNT13; GALNT15; GALNT7; Galntl2; mpp-GalNAc-T15; PIH5; polypeptide GalNAc transferase 15; polypeptide GalNAc transferase-like protein 2; polypeptide N-acetylgalactosaminyltransferase 13; polypeptide N-acetylgalactosaminyltransferase 15; polypeptide N-acetylgalactosaminyltransferase-like 2; polypeptide N-acetylgalactosaminyltransferase-like protein 2; pp-GalNAc-T15; pp-GaNTase-like protein 2; pregnancy-induced hypertension syndrome-related protein 5; protein-UDP acetylgalactosaminyltransferase-like protein 2; UDP-GalNAc transferase T15; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 7; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 2; UNQ770/PRO1564
Gene Symbols GALNT15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT15. Recombinant protein control fragment (Product #RP-90214).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.