missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT14, Mouse anti-Human, Clone: 2A8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00079623-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: KNGDDRQQWTKTGSHIEHIASHLCLDTDMFGDGTENGKEIVVNPCESSLMSQHWDMVSSpecifications
| GALNT14 | |
| Monoclonal | |
| Unconjugated | |
| UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14) | |
| FLJ12691/FLJ13977/GALNT15/GalNac-T10/GalNac-T14 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2A8 | |
| In 1x PBS, pH 7.4 | |
| NM_024572 | |
| GALNT14 | |
| GALNT14 (NP_078848.2, 494 a.a. ∼ 551 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 79623 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction