missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALNT13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543522
This item is not returnable.
View return policy
Description
Peptide sequence: CNKCDDKKER SLLPALRAVI SRNQEGPGEM GKAVLIPKDD QEKMKELFKI Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM].
Specifications
| GALNT13 | |
| Polyclonal | |
| Unconjugated | |
| GALNT13 | |
| A230002A12; A230020F20; BB182356; FLJ16031; FLJ41157; GalNAc transferase 13; GalNAc-T13; Galnt13; H_NH0187G20.1; KIAA1918; MGC119459; MGC119461; Polypeptide GalNAc transferase 13; polypeptide N-acetylgalactosaminyltransferase 13; pp-GalNAc-T13; pp-GaNTase 13; protein-UDP acetylgalactosaminyltransferase 13; T13; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 (GalNAc-T13); WUGSC:H_NH0187G20.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114805 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IUC8 | |
| GALNT13 | |
| Synthetic peptide directed towards the N-terminal of human GALNT13 (aa 31-80). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction