missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GALNT10 Polyclonal Antibody

Product Code. 12848932
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848932 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848932 Supplier Invitrogen™ Supplier No. PA520786

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20786 can be used with blocking peptide PEP-0900.

GALNT10 is responsible for encoding a GalNAc polypeptide N-acetylgalactosaminyltransferase enzyme, which plays a crucial role in the synthesis of mucin-type oligosaccharides. The enzyme transfers GalNAc from UDP-GalNAc to either serine or threonine residues of polypeptide acceptors. It has been observed that the protein encoded by this gene may exhibit higher catalytic activity towards glycosylated peptides as compared to non-glycosylated peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT10
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GALNT10
Gene Accession No. Q6P9S7, Q86SR1, Q925R7
Gene Alias AU018154; C330012K04Rik; GalNAc transferase 10; GALNACT10; galNAc-T10; GalNAc-T9; GALNT10; Galnt9; polypeptide GalNAc transferase 10; Polypeptide N-acetylgalactosaminyltransferase 10; PP PPGANTASE10; PPGALNACT10; ppGaNTase; pp-GaNTase 10; PPGANTASE10; protein-UDP acetylgalactosaminyltransferase 10; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 9; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10 (GalNAc-T10); UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 9
Gene Symbols GALNT10
Host Species Rabbit
Immunogen A 16 amino acid peptide near the amino terminus of human GALNT10.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170501, 171212, 55568
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.