missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14933-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes N-acetylgalactosamine-6-sulfatase whichis a lysosomal exohydrolase required for the degradation of theglycosaminoglycans, keratan sulfate, and chondroitin 6-sulfate.Sequence alterations including point, missense and nonsensemutations, as well as those that affect splicing, result in adeficiency of this enzyme. Deficiencies of this enzyme lead toMorquio A syndrome, a lysosomal storage disorder.Specifications
| GALNS | |
| Polyclonal | |
| Unconjugated | |
| GALNS | |
| Chondroitinase, Chondroitinsulfatase, Galactose 6 sulfate sulfatase, GALNAC6S, GalNAc6S sulfatase, GALNS, GAS, MPS4A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2588, 292073, 50917 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P34059, Q32KJ6, Q571E4 | |
| Galns | |
| GALNS Fusion Protein Ag6746 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction