Learn More
Invitrogen™ GALNS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522098
Description
Recommended positive controls: H1299, Neuro2A, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (89%), Rat (90%), Zebrafish (81%), Xenopus laevis (84%), Xenopus Tropicalis (80%), Dog (92%), Pig (90%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes N-acetylgalactosamine-6-sulfatase whichis a lysosomal exohydrolase required for the degradation of theglycosaminoglycans, keratan sulfate, and chondroitin 6-sulfate.Sequence alterations including point, missense and nonsensemutations, as well as those that affect splicing, result in adeficiency of this enzyme. Deficiencies of this enzyme lead toMorquio A syndrome, a lysosomal storage disorder.
Specifications
| GALNS | |
| Polyclonal | |
| Unconjugated | |
| GALNS | |
| Chondroitinase; chondroitinsulfatase; galactosamine (N-acetyl)-6-sulfatase; galactosamine (N-acetyl)-6-sulfate sulfatase; galactose-6-sulfate sulfatase; GalN6S; GALNAC6S; GalNAc6S sulfatase; Galns; GAS; mFLJ00319; MPS4A; N-acetylgalactosamine-6-sulfatase; N-acetylgalactosamine-6-sulfate sulfatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2588, 292073, 50917 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P34059, Q32KJ6, Q571E4 | |
| GALNS | |
| Recombinant protein encompassing a sequence within the center region of human GALNS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.