missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gallocyanine

Artikelnummer. 11479763
Ansicht ändern
Click to view available options
Quantity:
50g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479763 50g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11479763 Supplier Alfa Aesar™ Supplier No. A12936.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
Gallocyanine is used as an intermediate in chemical research. It is a small molecule dye that also inhibits DKK1 of the Wnt pathway.

Solubility
Soluble in water (10 mg/ml), 100% ethanol (10 mg/ml), alcohol, 1N NH4OH (10 mg/ml), and DMSO (25 mg/ml).

Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1562-85-2
Molecular Formula C15H13ClN2O5
MDL Number MFCD00011926
Quantity 50g
Beilstein 307454
Merck Index 14,4355
Synonym gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
Solubility Information Soluble in water (10mg/ml),100% ethanol (10mg/ml),alcohol,1N NH4OH (10mg/ml),and DMSO (25mg/ml).
InChI Key AQSOTOUQTVJNMY-UHFFFAOYSA-N
SMILES CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]
IUPAC Name 7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
Molecular Weight (g/mol) 336.728
PubChem CID 73801
Formula Weight 336.73
Chemical Name or Material Gallocyanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.