missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin gallate

Product Code. 15465049
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465049 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15465049 Supplier Thermo Scientific Alfa Aesar Supplier No. J62740.MA
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4233-96-9 | C22H18O11 | 458.375 g/mol

An antioxidant from green tea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4233-96-9
Color White
Molecular Formula C22H18O11
MDL Number MFCD00214298
Quantity 10 mg
Synonym =--gallocatechin gallate, gallocatechin gallate, =--gallocatechol gallate, unii-irw3c4y31q, gallocatechin gallate gcg, irw3c4y31q, gallocatechin gallate,-, =--gallocatechin-3-o-gallate, 2s,3r-5,7-dihydroxy-2-3,4,5-trihydroxyphenyl chroman-3-yl 3,4,5-trihydroxybenzoate, 2s,3r-2-3,4,5-trihydroxyphenyl-3,4-dihydro-1 2h-benzopyran-3,5,7-triol 3-3,4,5-trihydroxybenzoate
InChI Key WMBWREPUVVBILR-NQIIRXRSSA-N
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
IUPAC Name [(2S,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 458.375
PubChem CID 199472
Formula Weight 458.38
Physical Form Powder
Chemical Name or Material (-)-Gallocatechin gallate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.