missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin gallate, MedChemExpress

Product Code. p-200098966
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16469008

Brand: Medchem Express HYN0522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-Gallocatechin gallate is the polyphenol isolated from tea, with cancer-preventive activities.

(-)-Gallocatechin gallate is the polyphenol isolated from tea, with cancer-preventive activities.

Specifications

Chemical Name or Material (-)-Gallocatechin gallate
CAS 4233-96-9
Color White
Purity Grade Notes Research
Molecular Formula C22H18O11
Quantity 10 mg
Synonym (-)-Gallocatechol gallate
Solubility Information DMSO : ≥ 100 mg/mL (218.16 mM)
SMILES O=C(O[C@H]1[C@H](C2=CC(O)=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1)C4=CC(O)=C(O)C(O)=C4
Molecular Weight (g/mol) 458.37
Formula Weight 458.37
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.