missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin gallate, MedChemExpress

Product Code. 16469008
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469008 10 mg 10mg
16453876 25 mg 25mg
16451428 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16469008 Supplier Medchem Express Supplier No. HYN0522
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (-)-Gallocatechin gallate is the polyphenol isolated from tea, with cancer-preventive activities.

(-)-Gallocatechin gallate is the polyphenol isolated from tea, with cancer-preventive activities.

Specifications

Chemical Name or Material (-)-Gallocatechin gallate
CAS 4233-96-9
Color White
Purity Grade Notes Research
Molecular Formula C22H18O11
Quantity 10 mg
Synonym (-)-Gallocatechol gallate
Solubility Information DMSO : ≥ 100 mg/mL (218.16 mM)
SMILES O=C(O[C@H]1[C@H](C2=CC(O)=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1)C4=CC(O)=C(O)C(O)=C4
Molecular Weight (g/mol) 458.37
Formula Weight 458.37
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.