missing translation for 'onlineSavingsMsg'
Learn More

Gallium phosphide, 99.999% (metals basis)

Product Code. 11381609
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11381609

Brand: Thermo Scientific Alfa Aesar 012266.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Semiconducting Devices | CAS: 12063-98-8 | GaP | 100.70 g/mol

Gallium phosphide (GaP) is used in semiconductor devices, and in the manufacture of light-emitting diodes (LEDs) doped with other elements or in combination with gallium arsenide phosphide. With appropriate dopants, p-type and n-type semiconductors can be produced. It is used in optical systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12063-98-8
Molecular Formula GaP
Molecular Weight (g/mol) 100.70
MDL Number MFCD00016109
InChI Key HZXMRANICFIONG-UHFFFAOYSA-N
Synonym gallium phosphide, gallium monophosphide, unii-3j421f73dv, ccris 4019, gallium phosphide, polycryst, gallium phosphide trace metals basis, gallium phosphide trace metals basis 1g, gallium phosphide, single crystal substrate , <111>, diam. x thickness 2 in. x 0.5 mm
PubChem CID 82901
IUPAC Name gallanylidynephosphane
SMILES P#[Ga]

Specifications

Melting Point 1465°C
Color Orange
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 100.69
Density 4.13 g/cm3
Chemical Name or Material Gallium phosphide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.