missing translation for 'onlineSavingsMsg'
Learn More

Gallium pellets, 99.9999% (metals basis)

Product Code. 11408440
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11408440

Brand: Thermo Scientific Alfa Aesar 044324.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-55-3 | Ga | 69.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Quantity 10 g
Assay Percent Range 99.9999%, metal basis
Form Pellet
Chemical Name or Material Gallium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.