missing translation for 'onlineSavingsMsg'
Learn More

Gallium metal, packaged in poylethylene bottle, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11428620
Click to view available options
Quantity:
100 g
500 g
7440-55-3
Ga
69.72
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
5360835
CHEBI:49631
gallium
[Ga]
7440-55-3
Ga
69.72
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
5360835
CHEBI:49631
gallium
[Ga]

May be solid or liquid due to ambient temperature

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray
Density 5.91g/cm3
Assay Percent Range 99.9999% (Metals basis)
Quantity 100 g
Form Solid and/or Liquid
Chemical Name or Material Gallium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'