missing translation for 'onlineSavingsMsg'
Learn More

Gallium Indium Tin eutectic, 99.99% (metals basis)

Product Code. 11366948
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366948 50 g 50g
11376948 1 g 1g
11356948 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366948 Supplier Thermo Scientific Alfa Aesar Supplier No. 014634.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ga:In:Sn; 62:22:16 wt.%

Gallium-Indium eutectic (EGAIn) is an electrically conductive fluid metal. It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallium Indium Tin eutectic
Melting Point 10.7°C
Color White
Physical Form Liquid
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Molecular Formula Ga:In:Sn; 62:22:16 wt.%
MDL Number MFCD00144388
Solubility Information Insoluble in water or organic solvents.
Odor Odorless
Density 6.359
Show More Show Less
compliance-icons
Product Identifier
  • Gallium Indium Tin eutectic
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Gallium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.