missing translation for 'onlineSavingsMsg'
Learn More

Gallium Indium eutectic, 99.99% (metals basis)

Product Code. 11331819
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331819 10 g 10g
11341819 1 g 1g
11351819 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331819 Supplier Thermo Scientific Alfa Aesar Supplier No. 012478.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gallium Indium eutectic, Ga:In; 75.5:24.5 wt%, 10 g, 99.99% (metals basis), 6.2, Hazard Class: 8; Packaging Group: III, 231-163-8, H290-H314-H335, P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c, 15.7 deg.C, MFCD00144387, Liquid | Ga:In; 75.5:24.5 wt%

It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices. Gallium-Indium eutectic top contact, and a halogen light source was used to test P3HT-PCBM organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallium Indium eutectic
Melting Point 15.7°C
Density 6.2
Quantity 10 g
Assay Percent Range 99.99% (metals basis)
Form Liquid
Molecular Formula Ga:In; 75.5:24.5 wt%
MDL Number MFCD00144387
Solubility Information Insoluble in water or organic solvents.
compliance-icons
Product Identifier
  • Gallium Indium eutectic
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Gallium, Indium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.