missing translation for 'onlineSavingsMsg'
Learn More

Gallium Indium alloy, 99.99% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 044240.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11339768

  • £23.40 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Gallium Indium alloy
99.99% (metals basis)
MFCD00144387
Solid
Ga:In; 40:60 wt.%

Specifications

Gallium Indium alloy
1g
Solid
Ga:In; 40:60 wt.%
Silvery gray
Odorless
99.99% (metals basis)
MFCD00144387
Videos
SDS