missing translation for 'onlineSavingsMsg'
Learn More

Gallium Indium alloy, 99.99% (metals basis)

Product Code. 11329768
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329768 50 g 50g
11339768 1 g 1g
11319768 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329768 Supplier Thermo Scientific Alfa Aesar Supplier No. 044240.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ga:In; 40:60 wt.%

Step soldering involving temperature sensitive components, Soldering to molded interconnect device (MID) plastics, Fusible alloys/fuse applications, Mercury replacement, Thermal and electrical conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallium Indium alloy
Color Gray
Quantity 50 g
Odor Odorless
Assay Percent Range 99.99% (metals basis)
Form Solid
Molecular Formula Ga:In; 40:60 wt.%
MDL Number MFCD00144387
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.