missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. p-7051562
Click to view available options
Quantity:
0.2 g
1 g
5 g
Unit Size:
0.2g
1g
5g
This item is not returnable. View return policy

Product Code. 11304298

Brand: Thermo Scientific Alfa Aesar 041538.SS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Gallium(III) sulfide is used in semiconductor materials, functional ceramic materials, fluorescent powder and photoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-22-5
Molecular Formula Ga2S3
Molecular Weight (g/mol) 235.63
MDL Number MFCD00016106
InChI Key BVSHTEBQPBBCFT-UHFFFAOYSA-N
IUPAC Name digallium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Ga+3].[Ga+3]

Specifications

Melting Point 1255°C
Physical Form Powder
Quantity 0.2 g
UN Number UN3260
Solubility Information It dissolves in water slowly,which is easy to dissolve in concentrated alkaline to get gallate salts. It is also dissolves in HCL and nitric acid.
Formula Weight 235.63
Percent Purity 99.999%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.77 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.