missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11326178
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326178 50 g 50g
11316178 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326178 Supplier Thermo Scientific Alfa Aesar Supplier No. 011152.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13780-42-2 | Ga2O12S3 | 427.61 g/mol

Gallium(III) sulfate hydrate is used in the preparation of other gallium compounds such as gallium oxide. It is supported on silica gel act as a solid acid catalyst, which is employed as a heterogeneous catalyst for the esterification of p-hydroxybenzoic acid and heptanol to give n-heptyl p-hydroxybenzoate. On reacting with strong acid cation ion exchange resin, it exhibits excellent catalytic activity, which is used for synthesizing amyl hexanoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13780-42-2
Molecular Formula Ga2O12S3
Molecular Weight (g/mol) 427.61
MDL Number MFCD00149725
InChI Key UXLRWZBQRAWBQA-UHFFFAOYSA-H
Synonym gallium iii sulfate hydrate, gallium sulphate hydrate, 2ga.3so4.h2o, digallium 3+ hydrate trisulfate, gallium iii sulfate hydrate-ga puratrem, sulfuric acid, galliumsalt 3:2 , hydrate 8ci,9ci
PubChem CID 16213072
IUPAC Name digallium;trisulfate;hydrate
SMILES [Ga+3].[Ga+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 105°C to 110°C
Physical Form Crystalline Aggregates
Quantity 50 g
Solubility Information Slightly soluble in water and acid.
Formula Weight 427.62 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 3.860 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.