missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, Puratronic™, 99.999% (metals basis)

Product Code. 11376168
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376168 5 g 5g
11396168 25 g 25g
11306178 100 g 100g
11386168 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11376168 Supplier Thermo Scientific Alfa Aesar Supplier No. 011151.06
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

Used in vacuum deposition. | CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example, in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-21-4
Molecular Formula Ga2O3
Molecular Weight (g/mol) 187.44
MDL Number MFCD00011020
InChI Key AJNVQOSZGJRYEI-UHFFFAOYSA-N
IUPAC Name digallium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ga+3].[Ga+3]

Specifications

Melting Point 1900°C
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in acids. Insoluble in water
Formula Weight 187.44
Percent Purity 99.999%
Packaging Plastic bottle
Refractive Index 1.92
Specific Gravity 6.44
Density 6.44
Chemical Name or Material Gallium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.