missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, 99.995% (metals basis)

Product Code. 11303408
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303408 5 g 5g
11393398 1 g 1g
11313408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11303408 Supplier Thermo Scientific Alfa Aesar Supplier No. 036318.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition | CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

For vacuum deposition. In the production of catalysts. Used as part of the manufacturing of semiconductor devices. Has been shown to demonstrate catalytic properties and has also been used as an insulating barrier in tight junctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-21-4
Molecular Formula Ga2O3
Molecular Weight (g/mol) 187.44
MDL Number MFCD00011020
InChI Key AJNVQOSZGJRYEI-UHFFFAOYSA-N
IUPAC Name digallium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ga+3].[Ga+3]

Specifications

Melting Point 1900°C
Physical Form Sintered Lump
Quantity 5 g
Merck Index 14,4346
Solubility Information Insoluble in water. Soluble in most acids.
Formula Weight 187.44
Percent Purity 99.995%
Odor Odorless
Refractive Index 1.92
Density 6.44 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.