missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 11313408
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
12024-21-4
Ga2O3
187.44
MFCD00011020
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
[O--].[O--].[O--].[Ga+3].[Ga+3]
This item is not returnable. View return policy
12024-21-4
Ga2O3
187.44
MFCD00011020
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
[O--].[O--].[O--].[Ga+3].[Ga+3]

For vacuum deposition

For vacuum deposition. In the production of catalysts. Used as part of the manufacturing of semiconductor devices. Has been shown to demonstrate catalytic properties and has also been used as an insulating barrier in tight junctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1900°C
Physical Form Sintered Lump
Quantity 25 g
Merck Index 14,4346
Solubility Information Insoluble in water. Soluble in most acids.
Formula Weight 187.44
Percent Purity 99.995%
Odor Odorless
Refractive Index 1.92
Density 6.44 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'