missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number 10338100
Click to view available options
Quantity:
5 g
50 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
12024-21-4
Ga2O3
187.44
MFCD00011020
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
[O--].[O--].[O--].[Ga+3].[Ga+3]
This item is not returnable. View return policy
12024-21-4
Ga2O3
187.44
MFCD00011020
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
[O--].[O--].[O--].[Ga+3].[Ga+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 1740°C
Color White
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 99.99+%
Linear Formula Ga2O3
Fieser 04,241
Merck Index 15, 4382
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in acids,alkalies
Formula Weight 187.44
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 5.88
Density 5.8800g/mL
Chemical Name or Material Gallium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'