missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11348327
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
12024-21-4
Ga2O3
187.44
MFCD00011020
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
[O--].[O--].[O--].[Ga+3].[Ga+3]
This item is not returnable. View return policy
12024-21-4
Ga2O3
187.44
MFCD00011020
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
[O--].[O--].[O--].[Ga+3].[Ga+3]

In spectroscopic analysis

In spectroscopic analysisGallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. Used as an evaporated material and sputtering target of 99.999% in dielectric films. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1900°C
Physical Form Powder
Quantity 5 g
Merck Index 14,4346
Solubility Information Insoluble in water; Soluble in alkalies; Very slightly soluble in hot acids.
Formula Weight 187.44
Percent Purity 99.99%
Odor Odorless
Refractive Index 1.92
Density 6.44 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'