missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) nitride, 99.99% (metals basis)

Product Code. 11389367
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389367 50 g 50g
11399367 10 g 10g
11309377 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389367 Supplier Thermo Scientific Alfa Aesar Supplier No. 040218.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25617-97-4 | GaN | 83.73 g/mol

Gallium(III) nitride is used to read blu-ray discs. It is used in various wireless infrastructure applications. It finds applications in optoelectronic, high-power and high-frequency devices. It is also useful in military electronics (active electronically scanned array radars). It is biocompatible and used in electrodes and electronic components in living organisms. It is a suitable material for solar cell arrays for satellites. It is employed in bright light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25617-97-4
Molecular Formula GaN
Molecular Weight (g/mol) 83.73
MDL Number MFCD00016108
InChI Key JMASRVWKEDWRBT-UHFFFAOYSA-N
Synonym gallium nitride, gallium mononitride, unii-1r9cc3p9vl, gallium nitride,, 1r9cc3p9vl, gallium nitride trace metals basis, gallium iii nitride trace metals basis 10g
PubChem CID 117559
IUPAC Name azanylidynegallane
SMILES N#[Ga]

Specifications

Melting Point 800°C (sublimation)
Color Yellow
Physical Form Powder
Quantity 50 g
Merck Index 14,4351
Solubility Information Slightly soluble in hot concentrated sulfuric acid and hot conc; sodium hydroxide; Insoluble in water and dilute acids.
Formula Weight 83.73
Percent Purity 99.99%
Sensitivity Moisture sensitive
Density 6.1 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) nitride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.