missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11366168
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366168 100 g 100g
15425975 5 g 5g
11346168 25 g 25g
11356168 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11366168 Supplier Thermo Scientific Alfa Aesar Supplier No. 011150.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69365-72-6 | GaN3O9 | 255.74 g/mol

Gallium(III) nitrate hydrate is the potential precursor for the synthesis of other gallium compounds. It finds application in the preparation of gallium nitride, which is used as a solar cell array for satellites. In the field of pharmaceutical industry, it is used to treat symptomatic hypercalcemia by preventing the breakdown of bone by slowing osteoclast activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69365-72-6
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
Synonym gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis
PubChem CID 11550823
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 110°C (decomposition)
Color White
Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Merck Index 14,4346
Solubility Information Soluble in water.
IUPAC Name gallium;trinitrate;hydrate
Formula Weight 255.73 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Gallium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.