missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) fluoride, 99.995% (metals basis)

Product Code. p-7049317
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11351499

Brand: Thermo Scientific Alfa Aesar 013987.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-51-9

Gallium(III) fluoride is employed in the preparation and study of optical fluorogallate glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-51-9
MDL Number MFCD00011021

Specifications

Boiling Point ∼1000°C
Melting Point 800°C (sublimation)
Color White
Physical Form Powder
Quantity 1 g
UN Number UN3260
Merck Index 14,4354
Solubility Information Soluble in water (0.02g/L at 200C).
Formula Weight 126.72
Percent Purity 99.999%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.47 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.