missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) ethoxide

Product Code. 11396269
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396269 1 g 1g
11306279 5 g 5g
11316279 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396269 Supplier Thermo Scientific Alfa Aesar Supplier No. 044859.03
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2572-25-0 | C6H18GaO3 | 207.93 g/mol

Gallium(III) ethoxide cohydrolysis with Te alkoxides on PMMA yields heat-mode erasable optical memory. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2572-25-0
Molecular Formula C6H18GaO3
Molecular Weight (g/mol) 207.93
MDL Number MFCD01866730
InChI Key JWISYUBLQXUTOS-UHFFFAOYSA-N
Synonym gallium iii ethoxide
PubChem CID 131876769
IUPAC Name ethanol;gallium
SMILES [Ga].CCO.CCO.CCO

Specifications

Color Yellow
Density 1.25 g/mL
Flash Point 68°C (154°F)
Linear Formula Ga(OC2H5)3·xHOCH2CH3
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene.
Formula Weight 204.9
Physical Form Liquid
Chemical Name or Material Gallium (III) ethoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.