missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium(III) ethoxide, Thermo Scientific Chemicals
208.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Gallium(III) ethoxide cohydrolysis with Te alkoxides on PMMA yields heat-mode erasable optical memory. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Density | 1.25 g/mL |
Flash Point | 68°C (154°F) |
Linear Formula | Ga(OC2H5)3·xHOCH2CH3 |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in toluene. |
Formula Weight | 204.9 |
Physical Form | Liquid |
Chemical Name or Material | Gallium (III) ethoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'