missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific™

Product Code. 11368527
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368527 100 g 100g
11358527 5 g 5g
11378527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368527 Supplier Thermo Scientific Alfa Aesar Supplier No. 035698.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.073
MDL Number MFCD00011018
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl

Specifications

Boiling Point 201°C
Melting Point 78°C
Color White
Physical Form Beads
Quantity 100 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Very soluble in water. Soluble in benzene,carbon tetrachloride and carbon disulfide
Formula Weight 176.03
Sensitivity Moisture sensitive
Appearance Ultra dry. White powder/flake/crystalline/beads, etc.
Density 2.47
Chemical Name or Material Gallium(III) chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.