missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) chloride, ultra dry, 99.999% (metals basis)

Product Code. 11340558
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340558 100 g 100g
11330558 5 g 5g
11354908 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340558 Supplier Thermo Scientific Alfa Aesar Supplier No. 043879.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13450-90-3 | Cl3Ga | 176.073 g/mol

Gallium(III) chloride is used as lewis acid or precursor of organogallium reagents. It is used in detection of solar neutrinos. It is used in semiconductor devices for microelectronics and optics. It acts as a catalyst for a number of transformations such as the ring opening and recyclization of epoxide or trimerization of alkynes and as diamagnetic analogue for ferric compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.073
MDL Number MFCD00011018
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl

Specifications

Boiling Point 201°C
Melting Point 78°C
Color White
Physical Form Crystalline
Quantity 100 g
UN Number UN3260
Merck Index 14,4348
Solubility Information Soluble in water,benzene,carbon tetrachloride and carbon disulphide.
Formula Weight 176.03
Percent Purity 99.999%
Packaging Ampule
Sensitivity Moisture sensitive
Density 2.47 g/mL
Assay (metals basis)
Chemical Name or Material Gallium (III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.