missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) bromide, ultra dry, 99.998% (metals basis)

Product Code. 11343887
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343887 5 g 5g
11333887 1 g 1g
15416135 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343887 Supplier Thermo Scientific Alfa Aesar Supplier No. 035697.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13450-88-9 | Br3Ga | 309.435 g/mol

Gallium bromide is used as a catalyst in the alkylation of aromatic compounds by alkyl bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-88-9
Molecular Formula Br3Ga
Molecular Weight (g/mol) 309.435
MDL Number MFCD00016100
InChI Key SRVXDMYFQIODQI-UHFFFAOYSA-K
Synonym gallium tribromide, gallium bromide gabr3, gallium iii bromide, gallium bromide, gabr3, gallium bromide, gabr3, gallium iii bromide, anhydrous, gallium iii bromide, ultra dry, gallium iii bromide, anhydrous, granular
PubChem CID 83477
IUPAC Name tribromogallane
SMILES [Ga](Br)(Br)Br

Specifications

Boiling Point 279°C
Melting Point 122°C
Color White
Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 309.45
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.69 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.