missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) 2,4-pentanedionate, 99.99% (metals basis)

Product Code. 11369727
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11369727

Brand: Thermo Scientific Alfa Aesar 042086.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14405-43-7 | C15H21GaO6 | 367.05 g/mol

It is used in research on Ga-containing materials. Gallium oxide thin films can be created with atomic layer epitaxy (ALE) by combining gallium acetylacetonate with either water or ozone as the precursor. It can also be used for the synthesis of high purity gallium nitride nano-wires and nano-needles at low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14405-43-7
Molecular Formula C15H21GaO6
Molecular Weight (g/mol) 367.05
MDL Number MFCD00013492
InChI Key IMMRYVXLCAMRNK-UHFFFAOYSA-N
Synonym gallium iii acetylacetonate, gallium iii 2,4-pentanedionate
PubChem CID 71311502
IUPAC Name (Z)-4-[[(Z)-4-oxopent-2-en-2-yl]oxy-[(E)-4-oxopent-2-en-2-yl]oxygallanyl]oxypent-3-en-2-one
SMILES [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 196°C to 198°C (decomposition)
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 367.05
Percent Purity 99.99%
Odor Odorless
Density 1.42 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.