Learn More
Gallium arsenide, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1238°C |
Color | Gray |
Physical Form | Polycrystalline Pieces |
Quantity | 2 g |
Assay Percent Range | 99.999% (Metals basis) |
Solubility Information | Soluble in hydrochloric acid. Insoluble in water,ethanol,methanol and acetone. |
Formula Weight | 144.64 |
Odor | Odorless |
Density | 5.31 g/cm3 |
Chemical Name or Material | Gallium arsenide |
RUO – Research Use Only