missing translation for 'onlineSavingsMsg'
Learn More

Gallium arsenide, 99.999% (metals basis)

Product Code. 11339269
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339269 50 g 50g
11349269 2 g 2g
11359269 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11339269 Supplier Thermo Scientific Alfa Aesar Supplier No. 088458.18
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For Semiconductors (Transistors, Lasers, Solar Cells) | CAS: 1303-00-0 | AsGa | 144.645 g/mol

Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-00-0
Molecular Formula AsGa
Molecular Weight (g/mol) 144.645
MDL Number MFCD00011017
InChI Key JBRZTFJDHDCESZ-UHFFFAOYSA-N
Synonym gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick
PubChem CID 14770
IUPAC Name gallanylidynearsane
SMILES [Ga]#[As]

Specifications

Melting Point 1238°C
Color Gray
Physical Form Polycrystalline Pieces
Quantity 50 g
Odor Odorless
Assay Percent Range 99.999% (Metals basis)
Density 5.31
Solubility Information Soluble in hydrochloric acid. Insoluble in water,ethanol,methanol and acetone.
Formula Weight 144.64
Chemical Name or Material Gallium arsenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.