missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid, TRC

Product Code. 30135388
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Unit Size:
25g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135388 25 g 25g
30011606 5 g 5g
17983335 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135388 Supplier Toronto Research Chemicals Supplier No. TRCG18897825G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7 H6 O5
Molecular Weight (g/mol) 170.12
Synonym Gallic Acid, Benzoic acid, 3,4,5-trihydroxy-, Gallic acid (7CI,8CI), 3,4,5-Trihydroxybenzoic acid, 3,4,5-Hydroxybenzoic acid, A 1925-04, NSC 20103, NSC 674319
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)c1cc(O)c(O)c(O)c1

Specifications

InChI Formula InChI=1S/C7H6O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,8-10H,(H,11,12)
Formula Weight 170.0215
Percent Purity >95%
Quantity 25 g
Chemical Name or Material Gallic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.