missing translation for 'onlineSavingsMsg'
Learn More

Gallic acid monohydrate, ACS reagent

Product Code. p-4839143 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 13458618

Brand: Thermo Scientific Acros 447581000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 251.0°C
Linear Formula (HO)3C6H2CO2H·H2O
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, glycerol and acetone, practically insoluble in benzene, chloroform and, petroleum ether
IUPAC Name 3,4,5-trihydroxybenzoic acid;hydrate
Formula Weight 188.12
Grade ACS Reagent
Quantity 100 g
Packaging Plastic bottle
Chemical Name or Material Gallic acid monohydrate
compliance-icons
Product Identifier
  • Gallic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : gallic acid monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.