missing translation for 'onlineSavingsMsg'
Learn More

Gallic acid ethyl ester, 99%

Product Code. 10733275 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10733275 500 g Glass bottle 500g
10043510 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10733275 Supplier Thermo Scientific Acros Supplier No. 279935000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-61-8
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00016430
InChI Key VFPFQHQNJCMNBZ-UHFFFAOYSA-N
Synonym ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
PubChem CID 13250
ChEBI CHEBI:87247
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 151°C to 154°C
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 g
Beilstein 10, 484
Formula Weight 198.18
Percent Purity ≥98.5%
Physical Form Fine Powder
Chemical Name or Material Gallic acid ethyleester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.