missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gallery Reagents for Food and Beverage Analysis

Product Code. p-4529212 Shop All Thermo Scientific Products
Click to view available options
Type:
Sucrose System Reagent
Quantity:
20 mL Vials
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17861911

Brand: Thermo Scientific™ 984312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Choose from a continuously expanding selection of tests for automated food and beverage analysis.

Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for food and beverage analysis. Over 50 ready-to-use gallery system reagents are offered for food and beverage analysis, with the kits including two to four reagents in 20 mL barcoded vials. This enables the necessary identification data, such as lot numbers and expiration dates, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations. Save time and reduce errors by simplifying your analysis with the Gallery system reagents.

  • Kit structure includes 2 to 4 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, is automatically read
  • Photometric measurement is mainly based on the formation or consumption of NAD or NADH
  • Measurements are done at 340nm in 37°C
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations

Specifications

Type Sucrose System Reagent
Sample Type Food and Beverage Sample
Description Sucrose (Total Glucose) system reagents (300 tests)
Detects Sucrose (Total Glucose)
No. of Tests 300
Test Range 0.1 to 100 g/L, 15 to 500 mg/L
Analytical Method Enzymatic test with betafructosidase, hexokinase and glucose-6-phosphate dehydrogenase
Quantity 20 mL Vials
Interference Filter 340/ 600
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.